2-Mercaptopyridine-N-oxide sodium salt 40% in aqueous solution

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 2-Mercaptopyridinen-oxide sodiumsalt, 2-Mercaptopyridine N-oxide sodium

A16922.AE A16922.AP A16922.0F
ALFAA16922.AEEA 0 SGD
ALFAA16922.AE ALFAA16922.AP ALFAA16922.0F
2-Mercaptopyridine-N-oxide sodium salt 40% in aqueous solution
2-Mercaptopyridine N-oxide sodium salt

CAS No.: 3811-73-2 Molecular Formula: C5H4NNaOS Formula Weight: 149.15 Refractive Index: 1.4825 Beilstein Reference: 4026050 Merck Reference: 14,7994 MDL No.: MFCD01941547

Formula: C₅H₄NNaOS
MW: 149.15 g/mol
Density: 1.238
Storage Temperature: Ambient
MDL Number: MFCD01941547
CAS Number: 3811-73-2
EINECS: 223-296-5
Merck Index: 14,7994

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR