1-Bromo-4-fluoronaphthalene ≥98%

Supplier: Thermo Fisher Scientific
Warning

A16934.06 A16934.03 A16934.14
ALFAA16934.06EA 0 SGD
ALFAA16934.06 ALFAA16934.03 ALFAA16934.14
1-Bromo-4-fluoronaphthalene ≥98%
1-Bromo-4-fluoronaphthalene

Purity: 98% CAS No.: 341-41-3 Molecular Formula: C10H6BrF Formula Weight: 225.06 Flash Point: 110°(230°F) Melting Point: 33-37° Beilstein Reference: 2085804 MDL No.: MFCD00051473

Formula: C₁₀H₆BrF
MW: 225.06 g/mol
Melting Pt: 33…37 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00051473
CAS Number: 341-41-3
EINECS: 206-434-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR