N-Acetyl-L-tyrosine 99%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: N-Acetyl-L-Tyrosine, (S)-2-Acetamido-3-(4-hydroxyphenyl)propanoic acid, Ac-Tyr-OH

A17307.09 A17307.18 A17307.30
ALFAA17307.09EA 0 SGD
ALFAA17307.09 ALFAA17307.18 ALFAA17307.30
N-Acetyl-L-tyrosine 99%
N-Acetyl-L-tyrosine

Purity: 99% CAS No.: 537-55-3 Molecular Formula: C11H13NO4 Formula Weight: 223.23 Melting Point: 150-151° Beilstein Reference: 2697172 Optical Rotation: +58° (c=5 in ethanol) MDL No.: MFCD00037190

Formula: C₁₁H₁₃NO₄
Melting Pt: 150…151 °C
Storage Temperature: Ambient
MDL Number: MFCD00037190
CAS Number: 537-55-3
EINECS: 208-671-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR