2,6-Bis(hydroxymethyl)-p-cresol ≥90%, tech.

Supplier: Thermo Fisher Scientific
Warning

A17369.14 A17369.22 A17369.36
ALFAA17369.14EA 0 SGD
ALFAA17369.14 ALFAA17369.22 ALFAA17369.36
2,6-Bis(hydroxymethyl)-p-cresol ≥90%, tech.
2,6-Bis(hydroxymethyl)-p-cresol

Purity: 90% CAS No.: 91-04-3 Molecular Formula: C9H12O3 Formula Weight: 168.19 Melting Point: 119-126° Beilstein Reference: 1240237

Formula: C₉H₁₂O₃
Boiling Pt: 363 °C (1013 hPa)
Melting Pt: 119…126 °C
Density: 1,382 g/cm³ (–173 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004619
CAS Number: 91-04-3
EINECS: 202-036-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR