(±)-Ethyl mandelate 97

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Ethyl 2-hydroxy-2-phenylacetate

A17412.14 A17412.22
ALFAA17412.14EA 0 SGD
ALFAA17412.14 ALFAA17412.22
(±)-Ethyl mandelate 97
(±)-Ethyl mandelate

Purity: 97% CAS No.: 774-40-3 Molecular Formula: C10H12O3 Formula Weight: 180.20 Flash Point: 110°(230°F) Boiling Point: 253-255° MDL No.: MFCD00004494

Formula: C₁₀H₁₂O₃
MW: 180.2 g/mol
Boiling Pt: 253…255 °C
Density: 1.115
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004494
CAS Number: 774-40-3
EINECS: 212-263-0

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR