N,N-Diethyl-m-toluamide 97

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Diethyl-m-toluamide, Diethyltoluamide, N,N-Diethyl-3-methylbenzamide, Deet

A17486.0E A17486.22 A17486.36
ALFAA17486.0EEA 0 SGD
ALFAA17486.0E ALFAA17486.22 ALFAA17486.36
N,N-Diethyl-m-toluamide 97
N,N-Diethyl-m-toluamide

Purity: 97% CAS No.: 134-62-3 Molecular Formula: C12H17NO Formula Weight: 191.27 Flash Point: 145°(293°F) Refractive Index: 1.5230 Boiling Point: 110-112°/1mm Beilstein Reference: 2046711 Merck Reference: 14,2856 MDL No.: MFCD00009046

Formula: C₁₂H₁₇NO
MW: 191.27 g/mol
Boiling Pt: 110…112 °C (1 mmHg)
Melting Pt: –45 °C
Density: 0.997
Flash Pt: 145 °C (293 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009046
CAS Number: 134-62-3
EINECS: 205-149-7
Merck Index: 13,02876

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR