1,3-Bis(diethylamino)propane 97%

Supplier: Thermo Fisher Scientific
Danger

A17680.06 A17680.14
ALFAA17680.06EA 143 SGD
ALFAA17680.06 ALFAA17680.14
1,3-Bis(diethylamino)propane 97%
1,3-Bis(diethylamino)propane

Purity: 97% CAS No.: 60558-96-5 Molecular Formula: (CH3CH2)2N(CH2)3N(CH2CH3)2 Formula Weight: 186.34 Flash Point: 76°(168°F) Refractive Index: 1.4377 Boiling Point: 133°/80mm Beilstein Reference: 1741257 UN No.: 2735 MDL No.: MFCD00009057

Formula: C₁₁H₂₆N₂
Boiling Pt: 133 °C (80 mmHg)
Density: 0,810
Flash Pt: 76 °C (168 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009057
CAS Number: 60558-96-5
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR