Butyraldehyde ≥98%

Supplier: Thermo Scientific
Danger

Synonyms: Butylaldehyde, Butanal

A18243.0F
ALFAA18243.0FEA 0 SGD
ALFAA18243.0F
Butyraldehyde ≥98%
Butyraldehyde

Purity: 98+% CAS No.: 123-72-8 Molecular Formula: C4H8O Formula Weight: 72.11 Flash Point: -11°(12°F) Refractive Index: 1.3800 Odour: Pungent Melting Point: -99° Boiling Point: 75-76° Beilstein Reference: 506061 Merck Reference: 14,1591 UN No.: 1129 MDL No.: MFCD00007023

Formula: C₄H₈O
MW: 72,11 g/mol
Boiling Pt: 75…76 °C
Melting Pt: –99 °C
Density: 0,80
Flash Pt: –11 °C (–12 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007023
CAS Number: 123-72-8
EINECS: 204-646-6
UN: 1129
ADR: 3,II
Merck Index: 14,1591

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR