5-Chloro-2,1,3-benzothiadiazole 98%

Supplier: Thermo Fisher Scientific

Synonyms: 5-Chlorobenzo-2,1,3-thiadiazole

A18286.03 A18286.06
ALFAA18286.03EA 0 SGD
ALFAA18286.03 ALFAA18286.06
5-Chloro-2,1,3-benzothiadiazole 98%
5-Chloro-2,1,3-benzothiadiazole

Purity: 98% CAS No.: 2207-32-1 Molecular Formula: C6H3ClN2S Formula Weight: 170.62 Melting Point: 55-57° MDL No.: MFCD00174269

Formula: C₆H₃ClN₂S
MW: 170.62 g/mol
Melting Pt: 55…57 °C
Storage Temperature: Ambient
MDL Number: MFCD00174269
CAS Number: 2207-32-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR