2-Amino-6-bromobenzothiazole 98%

Supplier: Thermo Fisher Scientific
Warning

A18554.03 A18554.06 A18554.14
ALFAA18554.03EA 0 SGD
ALFAA18554.03 ALFAA18554.06 ALFAA18554.14
2-Amino-6-bromobenzothiazole 98%
2-Amino-6-bromobenzothiazole

Purity: 98% CAS No.: 15864-32-1 Molecular Formula: C7H5BrN2S Formula Weight: 229.10 Odour: Odourless

Formula: C₇H₅BrN₂S
MW: 229.1
Storage Temperature: Ambient
MDL Number: MFCD00152229
CAS Number: 15864-32-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR