Sodium alginate, very low viscosity

Supplier: Thermo Fisher Scientific

Synonyms: Alginic acid sodium salt, Algin

A18565.22 A18565.0E A18565.36
ALFAA18565.22EA 0 SGD
ALFAA18565.22 ALFAA18565.0E ALFAA18565.36
Sodium alginate, very low viscosity
Sodium alginate

CAS No.: 9005-38-3 Merck Reference: 14,241 MDL No.: MFCD00081310

Storage Temperature: Ambient
MDL Number: MFCD00081310
CAS Number: 9005-38-3
Merck Index: 14,241

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR