3-Bromo-4-chloroanisole ≥98%

Supplier: Thermo Fisher Scientific

A18660.03 A18660.06
ALFAA18660.03EA 0 SGD
ALFAA18660.03 ALFAA18660.06
3-Bromo-4-chloroanisole ≥98%
3-Bromo-4-chloroanisole

Purity: 98% CAS No.: 2732-80-1 Molecular Formula: C7H6BrClO Formula Weight: 221.48 MDL No.: MFCD00070739

Formula: C₇H₆BrClO
Storage Temperature: Ambient
MDL Number: MFCD00070739
CAS Number: 2732-80-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR