Ethyl methoxyacetate ≥98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Methoxyacetic ethyl ester

A18770.09 A18770.18
ALFAA18770.09EA 0 SGD
ALFAA18770.09 ALFAA18770.18
Ethyl methoxyacetate ≥98%
Ethyl methoxyacetate

Purity: 98% CAS No.: 3938-96-3 Molecular Formula: CH3OCH2CO2CH2CH3 Formula Weight: 118.13 Flash Point: 46°(115°F) Refractive Index: 1.401 Boiling Point: 44-46°/9mm Beilstein Reference: 1744761 UN No.: 3272 MDL No.: MFCD00009176

Formula: C₅H₁₀O₃
MW: 118.13 g/mol
Boiling Pt: 44…46 °C (9 mmHg)
Density: 1.00
Flash Pt: 46 °C (115 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009176
CAS Number: 3938-96-3
EINECS: 223-521-7
UN: 3272
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR