1-Bromo-2-methoxynaphthalene ≥97%

Supplier: Thermo Fisher Scientific
Warning

A18926.06 A18926.14 A18926.22
ALFAA18926.06EA 0 SGD
ALFAA18926.06 ALFAA18926.14 ALFAA18926.22
1-Bromo-2-methoxynaphthalene ≥97%
1-Bromo-2-methoxynaphthalene

Purity: 97% CAS No.: 3401-47-6 Molecular Formula: C11H9BrO Formula Weight: 237.10 Melting Point: 79-80° Beilstein Reference: 2045600

Formula: C₁₁H₉BrO
Melting Pt: 79…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00055374
CAS Number: 3401-47-6
EINECS: 222-272-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR