(R)-(-)-β-Aminophenethyl alcohol 98

Supplier: Thermo Fisher Scientific
Warning

Synonyms: D-Phenylglycinol, (R)-(-)-2-Phenylglycinol

A19030.03 A19030.06 A19030.14
ALFAA19030.03EA 0 SGD
ALFAA19030.03 ALFAA19030.06 ALFAA19030.14
(R)-(-)-β-Aminophenethyl alcohol 98
(R)-(-)-β-Aminophenethyl alcohol

Purity: 98% CAS No.: 56613-80-0 Molecular Formula: C8H11NO Formula Weight: 137.18 Melting Point: 76-78° Beilstein Reference: 2935848

Formula: C₈H₁₁NO
MW: 137.18
Melting Pt: 76…78 °C
Storage Temperature: Ambient
MDL Number: MFCD00008062
CAS Number: 56613-80-0
EINECS: 260-287-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR