2,5-Dibromoaniline 97%

Supplier: Thermo Fisher Scientific
Danger

A19114.06 A19114.14 A19114.22
ALFAA19114.06EA 95.5 SGD
ALFAA19114.06 ALFAA19114.14 ALFAA19114.22
2,5-Dibromoaniline 97%
2,5-Dibromoaniline

Purity: 98% CAS No.: 3638-73-1 Molecular Formula: C6H5Br2N Formula Weight: 250.93 Flash Point: 110°(230°F) Odour: Characteristic Beilstein Reference: 1817548 UN No.: 2811 MDL No.: MFCD00007636

Formula: C₆H₅Br₂N
MW: 250.92 g/mol
Boiling Pt: 281
Melting Pt: 52…56 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007636
CAS Number: 3638-73-1
EINECS: 222-865-5
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR