4-Bromo-3-fluorotoluene ≥98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 1-Bromo-2-fluoro-4-methylbenzene

A19142.09 A19142.14
ALFAA19142.09EA 0 SGD
ALFAA19142.09 ALFAA19142.14
4-Bromo-3-fluorotoluene ≥98%
4-Bromo-3-fluorotoluene

Purity: 98% CAS No.: 452-74-4 Molecular Formula: C7H6BrF Formula Weight: 189.03 Flash Point: 74°(165°F) Refractive Index: 1.5310 Boiling Point: 187-188° Beilstein Reference: 2500154 MDL No.: MFCD00040940

Formula: C₇H₆BrF
MW: 189.03 g/mol
Boiling Pt: 187…188 °C
Density: 1.494
Flash Pt: 74 °C (165 °F)
Storage Temperature: Ambient
MDL Number: MFCD00040940
CAS Number: 452-74-4
UN: 1993
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR