5-Chloro-2-nitro-p-toluidine ≥97%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 5-Chloro-4-methyl-2-nitroaniline

A19148.06
ALFAA19148.06EA 0 SGD
ALFAA19148.06
5-Chloro-2-nitro-p-toluidine ≥97%
5-Chloro-2-nitro-p-toluidine

Purity: 97% CAS No.: 7149-80-6 Molecular Formula: C7H7ClN2O2 Formula Weight: 186.60 Melting Point: 159-163° MDL No.: MFCD00034066

Formula: C₇H₇ClN₂O₂
MW: 186.60 g/mol
Melting Pt: 159…163 °C
Storage Temperature: Ambient
MDL Number: MFCD00034066
CAS Number: 7149-80-6
UN: 2237
ADR: 6.1 III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR