5-Bromo-o-anisaldehyde ≥98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 5-Bromo-2-methoxybenzaldehyde, 5-Bromo-2-anisaldehyde

A19396.14 A19396.22
ALFAA19396.14EA 0 SGD
ALFAA19396.14 ALFAA19396.22
5-Bromo-o-anisaldehyde ≥98%
5-Bromo-o-anisaldehyde

Purity: 98+% CAS No.: 25016-01-7 Molecular Formula: C8H7BrO2 Formula Weight: 215.05 Melting Point: 114-118° Beilstein Reference: 508840

Formula: C₈H₇BrO₂
MW: 215.05 g/mol
Boiling Pt: 269
Melting Pt: 114…118 °C
Storage Temperature: Ambient
MDL Number: MFCD00003347
CAS Number: 25016-01-7
EINECS: 246-564-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR