3-Bromo-4-hydroxybenzonitrile 98

Supplier: Thermo Fisher Scientific
Warning

A19400.06 A19400.14
ALFAA19400.06EA 0 SGD
ALFAA19400.06 ALFAA19400.14
3-Bromo-4-hydroxybenzonitrile 98
3-Bromo-4-hydroxybenzonitrile

Purity: 98% CAS No.: 2315-86-8 Molecular Formula: C7H4BrNO Formula Weight: 198.02 Melting Point: 156-161° Beilstein Reference: 2207020 UN No.: 3439 MDL No.: MFCD00143096

Formula: C₇H₄BrNO
MW: 198.02 g/mol
Boiling Pt: 406
Melting Pt: 156…161 °C
Storage Temperature: Ambient
MDL Number: MFCD00143096
CAS Number: 2315-86-8
EINECS: 219-022-9
UN: 3439
ADR: 6.1 III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR