2-Bromo-6-methoxynaphthalene ≥98%

Supplier: Thermo Fisher Scientific

A19450.09 A19450.18
ALFAA19450.09EA 0 SGD
ALFAA19450.09 ALFAA19450.18
2-Bromo-6-methoxynaphthalene ≥98%
2-Bromo-6-methoxynaphthalene

Purity: 98% CAS No.: 5111-65-9 Molecular Formula: C11H9BrO Formula Weight: 237.10 Melting Point: 105-110° Boiling Point: 160-164°/3mm MDL No.: MFCD00004062

Formula: C₁₁H₉BrO
Boiling Pt: 160…164 °C (3 mmHg)
Melting Pt: 105…110 °C
Storage Temperature: Ambient
MDL Number: MFCD00004062
CAS Number: 5111-65-9
EINECS: 225-837-0

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR