Benzyl cinnamate ≥99%

Supplier: Thermo Fisher Scientific

Synonyms: Cinnamic acid benzyl ester, trans-Cinnamic acid benzyl ester, 3-Phenyl-2-propenoic acid benzyl ester

A19550.22 A19550.36
ALFAA19550.22EA 0 SGD
ALFAA19550.22 ALFAA19550.36
Benzyl cinnamate ≥99%
Benzyl cinnamate

Purity: 99% CAS No.: 103-41-3 Molecular Formula: C16H14O2 Formula Weight: 238.29 Flash Point: 180°(356°F) Melting Point: 34-36° Boiling Point: 195-200°/5mm Beilstein Reference: 2051339 Merck Reference: 14,1130 MDL No.: MFCD00004789

Formula: C₁₆H₁₄O₂
MW: 238.29 g/mol
Boiling Pt: 195…200 °C (5 mmHg)
Melting Pt: 34…36 °C
Density: 1.11
Flash Pt: 180 °C (356 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004789
CAS Number: 103-41-3
EINECS: 203-109-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR