4-Amino-3-chlorophenol hydrochloride 98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 4-Amino-3-chlorophenol Hydrochloride, 2-Chloro-4-hydroxyaniline hydrochloride

A19720.06 A19720.14 A19720.22
ALFAA19720.06EA 0 SGD
ALFAA19720.06 ALFAA19720.14 ALFAA19720.22
4-Amino-3-chlorophenol hydrochloride 98%
4-Amino-3-chlorophenol hydrochloride

Purity: 98% CAS No.: 52671-64-4 Molecular Formula: C6H6ClNO.HCl Formula Weight: 180.04 Melting Point: 254-256° dec. MDL No.: MFCD00143110

Formula: C₆H₇Cl₂NO
Melting Pt: 254…256 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00143110
CAS Number: 52671-64-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR