Cyclohexanecarbonyl chloride ≥97%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Cyclohexanecarboxylic acid chloride, Cyclohexanecarboxylic chloride

A19824.14 A19824.22
ALFAA19824.14EA 0 SGD
ALFAA19824.14 ALFAA19824.22
Cyclohexanecarbonyl chloride ≥97%
Cyclohexanecarbonyl chloride

Purity: 97+% CAS No.: 2719-27-9 Molecular Formula: C7H11ClO Formula Weight: 146.62 Flash Point: 66°(150°F) Refractive Index: 1.4700 Boiling Point: 183-185° Beilstein Reference: 742163 UN No.: 3265 MDL No.: MFCD00001456

Formula: C₇H₁₁ClO
MW: 146.62 g/mol
Boiling Pt: 183…185 °C
Melting Pt: 29
Density: 1.088
Flash Pt: 66 °C (150 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001456
CAS Number: 2719-27-9
EINECS: 220-322-7
UN: 3265
ADR: 8 II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR