Ethyl benzofuroxan-5-carboxylate 97%

Supplier: Thermo Fisher Scientific

A19874.06
ALFAA19874.06EA 0 SGD
ALFAA19874.06
Ethyl benzofuroxan-5-carboxylate 97%
Ethyl benzofuroxan-5-carboxylate

Purity: 97% CAS No.: 17348-71-9 Molecular Formula: C9H8N2O3 Formula Weight: 208.18 Melting Point: 66-67° MDL No.: MFCD00276595

Formula: C₉H₈N₂O₃
MW: 208.18 g/mol
Melting Pt: 66…67 °C
Storage Temperature: Ambient
MDL Number: MFCD00276595
CAS Number: 17348-71-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR