Clorofene 96%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 2-Benzyl-4-chlorophenol

B20112.14
ALFAB20112.14EA 0 SGD
ALFAB20112.14
Clorofene 96%
Clorofene

Purity: 96% CAS No.: 120-32-1 Molecular Formula: C13H11ClO Formula Weight: 218.69 Flash Point: 185°(365°F) Melting Point: 45-51° Boiling Point: 160-162°/5mm Beilstein Reference: 1959194 Merck Reference: 14,2407 UN No.: 3077 MDL No.: MFCD00020140

Formula: C₁₃H₁₁ClO
MW: 218.68 g/mol
Boiling Pt: 160…162 °C (5 mmHg)
Melting Pt: 45…51 °C
Density: 1.190
Flash Pt: 185 °C (365 °F)
Storage Temperature: Ambient
MDL Number: MFCD00020140
CAS Number: 120-32-1
EINECS: 204-385-8
ADR: 9 III
Merck Index: 13,02433

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR