Tetrabromophenol blue

Supplier: Thermo Fisher Scientific

Synonyms: 4,5,6,7-Tetrabromo-3,3-bis(3,5-dibromo-4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide, 4,4'-(4,5,6,7-Tetrabromo-3H-2,1-benzoxathiol-3-ylidene)bis[2,6-dibromophenol] S,S-dioxide

B20123.03 B20123.06 B20123.14
ALFAB20123.03EA 0 SGD
ALFAB20123.03 ALFAB20123.06 ALFAB20123.14
Tetrabromophenol blue
Tetrabromophenol blue

CAS No.: 4430-25-5 Molecular Formula: C19H6Br8O5S Formula Weight: 985.55 Melting Point: 203-205° dec. Beilstein Reference: 378438 MDL No.: MFCD00005876

Formula: C₁₉H₆Br₈O₅S
Melting Pt: 203…205 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00005876
CAS Number: 4430-25-5
EINECS: 224-622-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR