Tetrabromophenol blue

Supplier: Thermo Fisher Scientific

B20123.03 B20123.06 B20123.14
ALFAB20123.03EA 0 SGD
ALFAB20123.03 ALFAB20123.06 ALFAB20123.14
Tetrabromophenol blue
Tetrabromophenol blue

CAS No.: 4430-25-5 Molecular Formula: C19H6Br8O5S Formula Weight: 985.55 Melting Point: 203-205° dec. Beilstein Reference: 378438 MDL No.: MFCD00005876

Formula: C₁₉H₆Br₈O₅S
Melting Pt: 203…205 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00005876
CAS Number: 4430-25-5
EINECS: 224-622-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR