3-Amino-4-bromo-5-methylisoxazole 97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 4-bromo-5-methylisoxazol-3-amine

B20138.03 B20138.06
ALFAB20138.03EA 0 SGD
ALFAB20138.03 ALFAB20138.06
3-Amino-4-bromo-5-methylisoxazole 97%
3-Amino-4-bromo-5-methylisoxazole

Purity: 97% CAS No.: 5819-40-9 Molecular Formula: C4H5BrN2O Formula Weight: 177.02 Melting Point: 73-75° MDL No.: MFCD00052553

Formula: C₄H₅BrN₂O
MW: 177.02 g/mol
Melting Pt: 73…75 °C
Storage Temperature: Ambient
MDL Number: MFCD00052553
CAS Number: 5819-40-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR