3-Ethyl-4-octanone 98%

Supplier: Thermo Fisher Scientific

B20175.06
ALFAB20175.06EA 0 SGD
ALFAB20175.06
3-Ethyl-4-octanone 98%
3-Ethyl-4-octanone

Purity: 98% CAS No.: 19781-29-4 Molecular Formula: C10H20O Formula Weight: 156.27 MDL No.: MFCD00048806

Formula: C₁₀H₂₀O
MW: 156.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00048806
CAS Number: 19781-29-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR