Mercaptoacetic acid (thioglycolic acid) ≥97%

Supplier: Thermo Fisher Scientific
Danger

B20391.0E B20391.22 B20391.36
ALFAB20391.0EEA 0 SGD
ALFAB20391.0E ALFAB20391.22 ALFAB20391.36
Mercaptoacetic acid (thioglycolic acid) ≥97%
Mercaptoacetic acid (Thioglycolic acid)

Purity: 97+% CAS No.: 68-11-1 Molecular Formula: C2H4O2S Formula Weight: 92.11 Flash Point: 119°(246°F) Physical Form: Clear liquid Density: 1.295 Refractive Index: 1.5045 Sensitivity: Air Sensitive Appearance: Colourless Odour: Stench Melting Point: -12° Boiling Point: 220° Beilstein Reference: 506166 Merck Reference: 14,9336 UN No.: 1940 MDL No.: MFCD00004876


Reagent that protects tryptophan in amino acid analysis

Formula: HSCH₂COOH
Boiling Pt: 220 °C
Melting Pt: –12 °C
Density: 1,295
Flash Pt: 119 °C (246 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004876
CAS Number: 68-11-1
EINECS: 200-677-4
UN: 1940
ADR: 8,II
Merck Index: 14,9336
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=45179825145ec44f2e058197a0ee54845ae7c750&vid=51e43f5b69b880b196570b3065f9b4d8db7336d1&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR