2-Mercaptopyridine-N-oxide sodium salt, anhydrous 98%

Supplier: Thermo Fisher Scientific
Warning

B20529.03 B20529.06 B20529.14
ALFAB20529.03EA 0 SGD
ALFAB20529.03 ALFAB20529.06 ALFAB20529.14
2-Mercaptopyridine-N-oxide sodium salt, anhydrous 98%
2-Mercaptopyridine N-oxide sodium salt

Purity: 98% CAS No.: 3811-73-2 Molecular Formula: C5H4NNaOS Formula Weight: 149.15 Melting Point: ca 290° dec. Merck Reference: 14,7994 MDL No.: MFCD01941547

Formula: C₅H₄NNaOS
Melting Pt: ∼ 290 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD01941547
CAS Number: 3811-73-2
EINECS: 223-296-5
Merck Index: 14,7994

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR