Ethyl trifluoropyruvate ≥97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Ethyl 3,3,3-trifluoro-2-oxopropanoate, Ethyl 3,3,3-trifluoropyruvate

B20611.06 B20611.14
ALFAB20611.06EA 0 SGD
ALFAB20611.06 ALFAB20611.14
Ethyl trifluoropyruvate ≥97%
Ethyl trifluoropyruvate

Purity: 97% CAS No.: 13081-18-0 Molecular Formula: C5H5F3O3 Formula Weight: 170.09 Flash Point: 27°(80°F) Refractive Index: 1.3405 Boiling Point: 95-98° Beilstein Reference: 2087388 UN No.: 3272 MDL No.: MFCD00114935

Formula: C₅H₅F₃O₃
MW: 170.09 g/mol
Boiling Pt: 95…98 °C
Density: 1.400
Flash Pt: 27 °C (80 °F)
Storage Temperature: Ambient
MDL Number: MFCD00114935
CAS Number: 13081-18-0
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR