α-Bromo-p-toluonitrile ≥98%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 4-(Bromomethyl)benzonitrile, 4-Cyanobenzyl bromide, p-(Bromomethyl)benzonitrile

B20641.06 B20641.14
ALFAB20641.06EA 0 SGD
ALFAB20641.06 ALFAB20641.14
α-Bromo-p-toluonitrile ≥98%
α-Bromo-p-toluonitrile

Purity: 98% CAS No.: 17201-43-3 Molecular Formula: C8H6BrN Formula Weight: 196.05 Melting Point: 113-117° Beilstein Reference: 636717 UN No.: 2923 MDL No.: MFCD00001829

Formula: C₈H₆BrN
MW: 196.05 g/mol
Boiling Pt: 135…137 °C (0.6 mmHg)
Melting Pt: 113…117 °C
Storage Temperature: Ambient
MDL Number: MFCD00001829
CAS Number: 17201-43-3
EINECS: 241-246-0
UN: 2923
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR