1-Pentanesulphonic acid sodium salt, Wasser_1 99% Dry weight

Supplier: Thermo Fisher Scientific

Synonyms: Sodium 1-pentanesulphonate, Sodium pentane-1-sulphonate, Pentane-1-sulphonic acid sodium salt, Sodium pentanesulphonate, 98%, Sodium pentanesulphonate

B20699.06 B20699.14 B20699.22
ALFAB20699.06EA 0 SGD
ALFAB20699.06 ALFAB20699.14 ALFAB20699.22
1-Pentanesulphonic acid sodium salt, Wasser_1 99% Dry weight
1-Pentanesulphonic acid sodium salt

Purity: 99% CAS No.: 22767-49-3 Molecular Formula: CH3(CH2)4SO3Na Formula Weight: 174.19 Melting Point: 300° Beilstein Reference: 3725147

Formula: C₅H₁₁O₃SNa
MW: 174.2 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007541
CAS Number: 22767-49-3
EINECS: 245-208-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ddb1c23c8a6fe3c156a3aacb62721e1d1a34efb0&vid=1936dcdb47ba374ce21fd758660f7ca6ba95d254&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR