p,α-Dimethylbenzyl alcohol ≥97%

Supplier: Thermo Fisher Scientific

Synonyms: 4-Methylphenyl methyl carbinol, 1-(p-Tolyl)ethanol, α,4-Dimethylbenzyl alcohol, 4-(1-Hydroxyethyl)toluene, 1-(4-Methylphenyl)ethanol

B20832.03 B20832.06 B20832.14
ALFAB20832.03EA 73.2 SGD
ALFAB20832.03 ALFAB20832.06 ALFAB20832.14
p,α-Dimethylbenzyl alcohol ≥97%
p,α-Dimethylbenzyl alcohol

Purity: 97% CAS No.: 536-50-5 Molecular Formula: C9H12O Formula Weight: 136.20 Flash Point: 107°(225°F) Refractive Index: 1.5230 Boiling Point: 218-220° Beilstein Reference: 1857562 Merck Reference: 14,3237 MDL No.: MFCD00016860

Formula: C₉H₁₂O
Boiling Pt: 218…220 °C
Melting Pt: 81…82 °C
Density: 0,987
Flash Pt: 107 °C (225 °F)
Storage Temperature: Ambient
MDL Number: MFCD00016860
CAS Number: 536-50-5
EINECS: 208-637-8
Merck Index: 14,3237

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR