Guanidinium thiocyanate ≥99%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Guanidinium sulphocyanide, GTC, Guanidine Thiocyanate, Guanidinium rhodanide

B21250.0E B21250.22 B21250.36
ALFAB21250.0EEA 0 SGD
ALFAB21250.0E ALFAB21250.22 ALFAB21250.36
Guanidinium thiocyanate ≥99%
Guanidinium thiocyanate

Purity: 99% CAS No.: 593-84-0 Molecular Formula: CH5N3.HSCN Formula Weight: 118.16 Melting Point: 118-122° Beilstein Reference: 3563461

Formula: NH₂C(=NH)NH₂·HSCN
MW: 118.16 g/mol
Melting Pt: 118…122 °C
Density: 1.29
Storage Temperature: Ambient
MDL Number: MFCD00013027
CAS Number: 593-84-0
EINECS: 209-812-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR