Ethyl-4-fluorobenzoate 99

Supplier: Thermo Fisher Scientific

Synonyms: 4-Fluorobenzoic acid ethyl ester, Ethyl 4-fluorobenzoate

B21484.06 B21484.14 B21484.22
ALFAB21484.06EA 0 SGD
ALFAB21484.06 ALFAB21484.14 ALFAB21484.22
Ethyl-4-fluorobenzoate 99
Ethyl 4-fluorobenzoate

Purity: 99% CAS No.: 451-46-7 Molecular Formula: C9H9FO2 Formula Weight: 168.17 Flash Point: 81°(177°F) Refractive Index: 1.4870 Odour: Recognizable Melting Point: 25-27° Boiling Point: 209-210° Beilstein Reference: 1866794 Merck Reference: 14,4171 MDL No.: MFCD00000351

Formula: C₉H₉FO₂
MW: 168.17 g/mol
Boiling Pt: 209…210 °C
Melting Pt: 25…27 °C
Density: 1.146
Flash Pt: 81 °C (177 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000351
CAS Number: 451-46-7
EINECS: 207-194-8
Merck Index: 13,04198

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR