Bis(diethylamino)phosphorochloridate 97%

Supplier: Thermo Fisher Scientific
Danger

B21519.14
ALFAB21519.14EA 0 SGD
ALFAB21519.14
Bis(diethylamino)phosphorochloridate 97%
Bis(diethylamino)phosphorochloridate

Purity: 97% CAS No.: 1794-24-7 Molecular Formula: [(CH3CH2)2N]2P(O)Cl Formula Weight: 226.69 Boiling Point: 101°/0.3mm UN No.: UN3265 MDL No.: MFCD00137958

Formula: [(CH₃CH₂)₂N]₂P(O)Cl
Boiling Pt: 101 °C (0,3 mmHg)
Density: 1,07
Storage Temperature: Ambient
MDL Number: MFCD00137958
CAS Number: 1794-24-7
EINECS: 217-262-9
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR