Ethyl-3-hydroxybutyrate ≥98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Ethyl 3-hydroxybutanoate, DL-Ethyl-3-hydroxybutyrate, Ethyl 3-hydroxybutyrate

B21521.14 B21521.22
ALFAB21521.14EA 59.5 SGD
ALFAB21521.14 ALFAB21521.22
Ethyl-3-hydroxybutyrate ≥98%
Ethyl 3-hydroxybutyrate

Purity: 98+% CAS No.: 5405-41-4 Molecular Formula: C6H12O3 Formula Weight: 132.16 Flash Point: 64°(148°F) Refractive Index: 1.4210 Boiling Point: 169-171° Beilstein Reference: 1446190 MDL No.: MFCD00004545

Formula: C₆H₁₂O₃
MW: 132.16 g/mol
Boiling Pt: 169…171 °C
Density: 1.013
Flash Pt: 64 °C (148 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004545
CAS Number: 5405-41-4
EINECS: 226-456-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR