Cyclohexyl hexanoate 99%

Supplier: Thermo Fisher Scientific

B21547.14 B21547.22
ALFAB21547.14EA 0 SGD
ALFAB21547.14 ALFAB21547.22
Cyclohexyl hexanoate 99%
Cyclohexyl hexanoate

Purity: 99% CAS No.: 6243-10-3 Molecular Formula: C12H22O2 Formula Weight: 198.31 MDL No.: MFCD00046356

Formula: C₁₂H₂₂O₂
MW: 198.31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00046356
CAS Number: 6243-10-3
EINECS: 228-354-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR