Ethyl bromofluoroacetate ≥97%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Bromofluoroacetic ethyl ester, Ethylbromofluoroacetate

B21579.06 B21579.14
ALFAB21579.06EA 0 SGD
ALFAB21579.06 ALFAB21579.14
Ethyl bromofluoroacetate ≥97%
Ethyl bromofluoroacetate

Purity: 97% CAS No.: 401-55-8 Molecular Formula: FBrCHCO2CH2CH3 Formula Weight: 184.99 Flash Point: 70°(158°F) Refractive Index: 1.4255 Boiling Point: 154° UN No.: 2810 MDL No.: MFCD00042095

Formula: BrFCHCOOCH₂CH₃
MW: 184.99 g/mol
Boiling Pt: 154 °C
Density: 1.540
Flash Pt: 70 °C (158 °F)
Storage Temperature: Ambient
MDL Number: MFCD00042095
CAS Number: 401-55-8
EINECS: 206-928-4
UN: 2810
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR