4-Bromo-2-methylanisole 98

Supplier: Thermo Fisher Scientific

Synonyms: 5-Bromo-2-methoxytoluene, 1-Bromo-4-methoxy-3-methylbenzene, 4-Bromo-1-methoxy-2-methylbenzene

B21621.09 B21621.18
ALFAB21621.09EA 0 SGD
ALFAB21621.09 ALFAB21621.18
4-Bromo-2-methylanisole 98
4-Bromo-2-methylanisole

Purity: 98+% CAS No.: 14804-31-0 Molecular Formula: C8H9BrO Formula Weight: 201.06 Melting Point: 66-69° Boiling Point: 124-125°/25mm MDL No.: MFCD01321139

Formula: C₈H₉BrO
MW: 201.06 g/mol
Boiling Pt: 124…125 °C (25 mmHg)
Melting Pt: 66…69 °C
Storage Temperature: Ambient
MDL Number: MFCD01321139
CAS Number: 14804-31-0

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR