Methyl-4-acetylbenzoate ≥99%

Supplier: Thermo Fisher Scientific

Synonyms: Methyl 4-acetylbenzoate

B21626.04 B21626.09
ALFAB21626.04EA 0 SGD
ALFAB21626.04 ALFAB21626.09
Methyl-4-acetylbenzoate ≥99%
Methyl 4-acetylbenzoate

Purity: 99% CAS No.: 3609-53-8 Molecular Formula: C10H10O3 Formula Weight: 178.19 Melting Point: 92-95° MDL No.: MFCD00216474

Formula: C₁₀H₁₀O₃
Melting Pt: 92…95 °C
Storage Temperature: Ambient
MDL Number: MFCD00216474
CAS Number: 3609-53-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR