2-Bromo-6-nitrotoluene ≥98%

Supplier: Thermo Fisher Scientific
Warning

B21658.06 B21658.14
ALFAB21658.06EA 0 SGD
ALFAB21658.06 ALFAB21658.14
2-Bromo-6-nitrotoluene ≥98%
2-Bromo-6-nitrotoluene

Purity: 98% CAS No.: 55289-35-5 Molecular Formula: C7H6BrNO2 Formula Weight: 216.04 Flash Point: 110°(230°F) Melting Point: 37-42° Boiling Point: 108-110°/3.3mm Beilstein Reference: 2502581 MDL No.: MFCD00009792

Formula: C₇H₆BrNO₂
Boiling Pt: 108…110 °C (3,3 mmHg)
Melting Pt: 37…42 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009792
CAS Number: 55289-35-5
EINECS: 259-566-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR