α-Bromo-o-xylene ≥98%

Supplier: Thermo Fisher Scientific
Danger

B21717.14 B21717.22
ALFAB21717.14EA 0 SGD
ALFAB21717.14 ALFAB21717.22
α-Bromo-o-xylene ≥98%
α-Bromo-o-xylene

Purity: 98% CAS No.: 89-92-9 Molecular Formula: C8H9Br Formula Weight: 185.07 Storage Temperature: 2°C to 8°C Flash Point: 82°(179°F) Refractive Index: 1.5750 Melting Point: 18-20° Boiling Point: 216-217° Beilstein Reference: 636333 Merck Reference: 14,10090 UN No.: 1701 MDL No.: MFCD00000174

Formula: C₈H₉Br
Boiling Pt: 216…217 °C
Melting Pt: 18…20 °C
Density: 1,381
Flash Pt: 82 °C (179 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00000174
CAS Number: 89-92-9
EINECS: 201-951-6
UN: 1701
ADR: 6.1,II
Merck Index: 14,10090

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR