Sodium tetradecyl sulphate 95% cont. up to ca 5% sodium methyl sulfate

Supplier: Thermo Fisher Scientific
Warning

B21941.03 B21941.06 B21941.14
ALFAB21941.03EA 0 SGD
ALFAB21941.03 ALFAB21941.06 ALFAB21941.14
Sodium tetradecyl sulphate 95% cont. up to ca 5% sodium methyl sulfate
Sodium tetradecyl sulphate

Purity: 95% CAS No.: 1191-50-0 Molecular Formula: CH3(CH2)13OSO3Na Formula Weight: 316.44 Melting Point: 182-192° Beilstein Reference: 3578122

Formula: C₁₄H₂₉NaO₄S
Melting Pt: 182…192 °C
Storage Temperature: Ambient
MDL Number: MFCD00007468
CAS Number: 139-88-8
EINECS: 214-737-2
UN: 3267
ADR: 8,III
Merck Index: 12,08838

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR