Trimethylsilyl acetate (acetoxytrimethylsilane) ≥97%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Trimethylsilyl acetate

B22037.14 B22037.22
ALFAB22037.14EA 0 SGD
ALFAB22037.14 ALFAB22037.22
Trimethylsilyl acetate (acetoxytrimethylsilane) ≥97%
Trimethylsilyl acetate (Acetoxytrimethylsilane)

Purity: 97% CAS No.: 2754-27-0 Molecular Formula: C5H12O2Si Formula Weight: 132.24 Flash Point: 4°(39°F) Refractive Index: 1.3880 Melting Point: -32° Boiling Point: 107° Beilstein Reference: 1744463 UN No.: 3272 MDL No.: MFCD00008695

Formula: C₅H₁₂O₂Si
MW: 132.24 g/mol
Boiling Pt: 107 °C
Melting Pt: –32 °C
Density: 0.884
Flash Pt: 4 °C (39 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008695
CAS Number: 2754-27-0
EINECS: 220-404-2
UN: 3272
ADR: 3 II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR