3'-Bromo-4'-fluoroacetophenone ≥98%

Supplier: Thermo Fisher Scientific
Warning

B22091.14 B22091.06
ALFAB22091.14EA 0 SGD
ALFAB22091.14 ALFAB22091.06
3'-Bromo-4'-fluoroacetophenone ≥98%
3'-Bromo-4'-fluoroacetophenone

Purity: 98+% CAS No.: 1007-15-4 Molecular Formula: C8H6BrFO Formula Weight: 217.04 Flash Point: 110°(230°F) Melting Point: 55-57° Boiling Point: 150°/12mm Beilstein Reference: 3088698 MDL No.: MFCD00042466

Formula: C₈H₆BrFO
MW: 217.04 g/mol
Boiling Pt: 150 °C (12 mmHg)
Melting Pt: 55…57 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00042466
CAS Number: 1007-15-4
EINECS: 213-750-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR