4-Fluoro-α-methylbenzyl alcohol ≥98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: p-Fluoro-α-methylbenzyl alcohol, 1-(4-Fluorophenyl)ethyl Alcohol, 1-(4-Fluorophenyl)ethanol, 4-Fluorophenyl methyl carbinol

B22097.14
ALFAB22097.14EA 0 SGD
ALFAB22097.14
4-Fluoro-α-methylbenzyl alcohol ≥98%
4-Fluoro-α-methylbenzyl alcohol

Purity: 98% CAS No.: 403-41-8 Molecular Formula: C8H9FO Formula Weight: 140.16 Storage Temperature: 2°C to 8°C Flash Point: 91°(196°F) Boiling Point: 90-92°/7mm Beilstein Reference: 2042025 MDL No.: MFCD00004515

Formula: C₈H₉FO
Boiling Pt: 90…92 °C (7 mmHg)
Melting Pt: 9,3 °C
Density: 1,109
Flash Pt: 91 °C (196 °F)
MDL Number: MFCD00004515
CAS Number: 403-41-8
EINECS: 206-959-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR